Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACCTTCAGCTTCTCGGCGTCCGA[C/G]TCCTGCTTCTTCTTGGCCGACGCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600106 MIM: 605955 | ||||||||||||||||||||
Literature Links: |
INPP5A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
INPP5A - inositol polyphosphate-5-phosphatase A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NKX6-2 - NK6 homeobox 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_177400.2 | 736 | Missense Mutation | GAC,GAG | D,E 219 | NP_796374.1 | |
XM_017016789.1 | 736 | Missense Mutation | GAC,GAG | D,E 219 | XP_016872278.1 |