Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATTATATACTTGTTTCTAAACTGT[C/T]ATCCAGTACTGTTAGCAGATACTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610000 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CEP55 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CEP55 - centrosomal protein 55 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127182.1 | Intron | NP_001120654.1 | ||||
NM_018131.4 | Intron | NP_060601.3 | ||||
XM_011539918.1 | Intron | XP_011538220.1 | ||||
XM_011539919.1 | Intron | XP_011538221.1 | ||||
XM_011539920.2 | Intron | XP_011538222.1 | ||||
XM_017016372.1 | Intron | XP_016871861.1 | ||||
XM_017016373.1 | Intron | XP_016871862.1 |