Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGTGAGATTTCTGGCTGACTTGGA[C/G]GCGCTCTGGGCCCGCTGAACAATGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613597 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HOGA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HOGA1 - 4-hydroxy-2-oxoglutarate aldolase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MORN4 - MORN repeat containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098831.1 | 716 | Intron | NP_001092301.1 | |||
NM_178832.3 | 716 | Silent Mutation | GCC,GCG | A,A 137 | NP_849154.1 | |
XM_011539250.2 | 716 | Silent Mutation | GCC,GCG | A,A 195 | XP_011537552.1 | |
XM_011539251.2 | 716 | Silent Mutation | GCC,GCG | A,A 137 | XP_011537553.1 |