Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGCCCTGATCTACCTGGTTCTCAT[C/T]GCTGTGGGGCAGAACTACATGAAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611815 MIM: 602669 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ELOVL3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ELOVL3 - ELOVL fatty acid elongase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152310.2 | 1405 | Silent Mutation | ATC,ATT | I,I 48 | NP_689523.1 | |
XM_011540245.2 | 1405 | Silent Mutation | ATC,ATT | I,I 48 | XP_011538547.1 |
PITX3 - paired like homeodomain 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |