Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGAGAGTCCCATGGGGTCTGGGA[C/T]GCAGCAAGAGCACAGTGGGCCCCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606701 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DRGX PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DRGX - dorsal root ganglia homeobox | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001276451.1 | 736 | Missense Mutation | ATC,GTC | I,V 184 | NP_001263380.1 | |
XM_011540089.2 | 736 | Missense Mutation | ATC,GTC | I,V 219 | XP_011538391.1 |