Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCATGTCAAACTTGATGAGCGGCGG[A/G]GCGATCACGCGCACCTCGGCGTTGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
LRRC20 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LRRC20 - leucine rich repeat containing 20 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278211.1 | 652 | Silent Mutation | GCC,GCT | A,A 164 | NP_001265140.1 | |
NM_001278212.1 | 652 | Silent Mutation | GCC,GCT | A,A 164 | NP_001265141.1 | |
NM_001278213.1 | 652 | Silent Mutation | GCC,GCT | A,A 114 | NP_001265142.1 | |
NM_001278214.1 | 652 | Silent Mutation | GCC,GCT | A,A 95 | NP_001265143.1 | |
NM_018205.3 | 652 | Silent Mutation | GCC,GCT | A,A 108 | NP_060675.1 | |
NM_018239.3 | 652 | Silent Mutation | GCC,GCT | A,A 114 | NP_060709.2 | |
NM_207119.2 | 652 | Silent Mutation | GCC,GCT | A,A 164 | NP_997002.1 | |
XM_017016374.1 | 652 | Silent Mutation | GCC,GCT | A,A 164 | XP_016871863.1 | |
XM_017016375.1 | 652 | Silent Mutation | GCC,GCT | A,A 164 | XP_016871864.1 |