Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTTCAGCTTCAATGTGCCCTTCAT[C/T]CCAGTCACCCAGGCCTCCGCTTCTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
SEC23IP PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SEC23IP - SEC23 interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007190.3 | 167 | Silent Mutation | ATC,ATT | I,I 38 | NP_009121.1 | |
XM_005269469.2 | 167 | Silent Mutation | ATC,ATT | I,I 38 | XP_005269526.1 | |
XM_005269470.2 | 167 | UTR 5 | XP_005269527.1 | |||
XM_017015610.1 | 167 | UTR 5 | XP_016871099.1 |