Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCTGGGGTCGAGTCGGCTGCTGG[A/G]GTTTATCATCTGGAAGATTCTCTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609177 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZWINT PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZWINT - ZW10 interacting kinetochore protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005413.1 | 756 | Missense Mutation | CCC,CTC | P,L 193 | NP_001005413.1 | |
NM_007057.3 | 756 | Missense Mutation | CCC,CTC | P,L 240 | NP_008988.2 | |
NM_032997.2 | 756 | Missense Mutation | CCC,CTC | P,L 240 | NP_127490.1 | |
XM_017015605.1 | 756 | Missense Mutation | CCC,TCC | P,S 228 | XP_016871094.1 |