Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACGATGTGTATAGTACAGTTTGAC[A/G]AGCCTGGGTCCATACAGACCGCTGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609230 | ||||||||||||||||||||
Literature Links: |
NUDT5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NUDT5 - nudix hydrolase 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321647.1 | 823 | Missense Mutation | TCG,TTG | S,L 136 | NP_001308576.1 | |
NM_001321648.1 | 823 | Missense Mutation | TCG,TTG | S,L 50 | NP_001308577.1 | |
NM_014142.3 | 823 | Missense Mutation | TCG,TTG | S,L 136 | NP_054861.2 |
SEC61A2 - Sec61 translocon alpha 2 subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |