Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCTTTCTCAATGTTCTCGTTCAG[A/G]TGACAATTCTTCTTGGTCTCAGCTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604055 MIM: 615391 | ||||||||||||||||||||
Literature Links: |
IDI1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IDI1 - isopentenyl-diphosphate delta isomerase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317955.1 | 1501 | Silent Mutation | CAC,CAT | H,H 41 | NP_001304884.1 | |
NM_001317956.1 | 1501 | Silent Mutation | CAC,CAT | H,H 41 | NP_001304885.1 | |
NM_001317957.1 | 1501 | Intron | NP_001304886.1 | |||
NM_004508.3 | 1501 | Silent Mutation | CAC,CAT | H,H 97 | NP_004499.2 | |
XM_011519457.1 | 1501 | Silent Mutation | CAC,CAT | H,H 41 | XP_011517759.1 | |
XM_017016191.1 | 1501 | Intron | XP_016871680.1 |
IDI2-AS1 - IDI2 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |