Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCAGAGATGTCTTCCAGAAGTAC[C/G]AAAGATTTAATTAAAAGTAAGTGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610000 | ||||||||||||||||||||
Literature Links: |
CEP55 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CEP55 - centrosomal protein 55 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127182.1 | 325 | Silent Mutation | ACC,ACG | T,T 6 | NP_001120654.1 | |
NM_018131.4 | 325 | Silent Mutation | ACC,ACG | T,T 6 | NP_060601.3 | |
XM_011539918.1 | 325 | UTR 5 | XP_011538220.1 | |||
XM_011539919.1 | 325 | UTR 5 | XP_011538221.1 | |||
XM_011539920.2 | 325 | UTR 5 | XP_011538222.1 | |||
XM_017016372.1 | 325 | UTR 5 | XP_016871861.1 | |||
XM_017016373.1 | 325 | UTR 5 | XP_016871862.1 |