Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTAAGCAGCCAAACTCGTACTGATA[C/T]CACCATGCAGCCAAGGTTACTGAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601107 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABCC2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ABCC2 - ATP binding cassette subfamily C member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000392.4 | Intron | NP_000383.1 | ||||
XM_006717630.3 | Intron | XP_006717693.1 | ||||
XM_006717631.3 | Intron | XP_006717694.1 | ||||
XM_011539291.2 | Intron | XP_011537593.1 | ||||
XM_017015675.1 | Intron | XP_016871164.1 |