Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGCTGCAGATATATGTGGAGTAA[C/G]TGCAGAACATTATGCTGTTACTTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610856 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKRD30A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANKRD30A - ankyrin repeat domain 30A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_052997.2 | 753 | Missense Mutation | ACT,AGT | T,S 184 | NP_443723.2 | |
XM_011519757.2 | 753 | Missense Mutation | ACT,AGT | T,S 240 | XP_011518059.1 | |
XM_011519758.2 | 753 | Intron | XP_011518060.1 | |||
XM_017016883.1 | 753 | Intron | XP_016872372.1 |