Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGGATAGGCCTGGAAATCTCAGAG[A/G]TTATCAGATGCCTGAGTGGGGAGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610659 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GRID1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GRID1 - glutamate ionotropic receptor delta type subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017551.2 | 4541 | UTR 3 | NP_060021.1 | |||
XM_011539720.2 | 4541 | Intron | XP_011538022.1 |
GRID1-AS1 - GRID1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |