Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAATTAGAAATGCTTTAATTTTTGC[A/C]AAGCGCTAAGTATAGTGCGTGCCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
SEC23IP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SEC23IP - SEC23 interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007190.3 | Intron | NP_009121.1 | ||||
XM_005269469.2 | Intron | XP_005269526.1 | ||||
XM_005269470.2 | Intron | XP_005269527.1 | ||||
XM_017015610.1 | Intron | XP_016871099.1 |