Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTATCTGTGGTCATACATTCCGGA[A/G]AGACAGTAGCTGAGGTCTCTCCCGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601037 | ||||||||||||||||||||
Literature Links: |
HNRNPF PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HNRNPF - heterogeneous nuclear ribonucleoprotein F | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098204.1 | 1212 | Silent Mutation | CTC,CTT | L,L 268 | NP_001091674.1 | |
NM_001098205.1 | 1212 | Silent Mutation | CTC,CTT | L,L 268 | NP_001091675.1 | |
NM_001098206.1 | 1212 | Silent Mutation | CTC,CTT | L,L 268 | NP_001091676.1 | |
NM_001098207.1 | 1212 | Silent Mutation | CTC,CTT | L,L 268 | NP_001091677.1 | |
NM_001098208.1 | 1212 | Silent Mutation | CTC,CTT | L,L 268 | NP_001091678.1 | |
NM_004966.3 | 1212 | Silent Mutation | CTC,CTT | L,L 268 | NP_004957.1 |