Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCACTTTTCTCTGTTAAAAATCAC[A/G]AAGTCTTGCTGGATGGCATCGAGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608407 | ||||||||||||||||||||
Literature Links: |
DPYSL4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DPYSL4 - dihydropyrimidinase like 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STK32C - serine/threonine kinase 32C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318878.1 | 1510 | Silent Mutation | TTC,TTT | F,F 446 | NP_001305807.1 | |
NM_001318879.1 | 1510 | Silent Mutation | TTC,TTT | F,F 316 | NP_001305808.1 | |
NM_001318880.1 | 1510 | Intron | NP_001305809.1 | |||
NM_001318881.1 | 1510 | Intron | NP_001305810.1 | |||
NM_001318882.1 | 1510 | Intron | NP_001305811.1 | |||
NM_001318883.1 | 1510 | Intron | NP_001305812.1 | |||
NM_173575.3 | 1510 | Silent Mutation | TTC,TTT | F,F 433 | NP_775846.2 | |
XM_011539688.1 | 1510 | Silent Mutation | TTC,TTT | F,F 451 | XP_011537990.1 | |
XM_011539690.2 | 1510 | Silent Mutation | TTC,TTT | F,F 438 | XP_011537992.1 | |
XM_011539693.2 | 1510 | Silent Mutation | TTC,TTT | F,F 316 | XP_011537995.1 | |
XM_011539694.1 | 1510 | Silent Mutation | TTC,TTT | F,F 316 | XP_011537996.1 | |
XM_011539695.1 | 1510 | Silent Mutation | TTC,TTT | F,F 316 | XP_011537997.1 | |
XM_011539696.1 | 1510 | Silent Mutation | TTC,TTT | F,F 316 | XP_011537998.1 | |
XM_017016100.1 | 1510 | Intron | XP_016871589.1 | |||
XM_017016101.1 | 1510 | Intron | XP_016871590.1 |