Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGTCCTTTCTTTTTTGAACAAGT[C/T]GGAACCTCTGATCCCGGCCGTATTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
TCERG1L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TCERG1L - transcription elongation regulator 1 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_174937.3 | 1741 | Missense Mutation | CAA,CGA | Q,R 552 | NP_777597.2 | |
XM_011539577.1 | 1741 | Missense Mutation | CAA,CGA | Q,R 565 | XP_011537879.1 | |
XM_011539578.2 | 1741 | Intron | XP_011537880.1 |
TCERG1L-AS1 - TCERG1L antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |