Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGTTGGGTGAATATTTCCAGGAAG[C/G]GGGCAGAGACGGGGTAGGAGAAGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605603 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MYOZ1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MYOZ1 - myozenin 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SYNPO2L - synaptopodin 2 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001114133.2 | 1705 | Missense Mutation | CCT,GCT | P,A 797 | NP_001107605.1 | |
NM_024875.4 | 1705 | Missense Mutation | CCT,GCT | P,A 573 | NP_079151.2 | |
XM_005270158.2 | 1705 | Missense Mutation | CCT,GCT | P,A 501 | XP_005270215.1 | |
XM_005270159.3 | 1705 | Missense Mutation | CCT,GCT | P,A 501 | XP_005270216.1 |