Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTTCGTGTGTTTCAAAGTCCAAGT[A/C]ATCATCACTATCTAAACATTCAAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616464 MIM: 609233 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ECD PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ECD - ecdysoneless cell cycle regulator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135752.1 | 1903 | Missense Mutation | GAC,TAC | D,Y 554 | NP_001129224.1 | |
NM_001135753.1 | 1903 | Missense Mutation | GAC,TAC | D,Y 478 | NP_001129225.1 | |
NM_007265.2 | 1903 | Missense Mutation | GAC,TAC | D,Y 521 | NP_009196.1 |
NUDT13 - nudix hydrolase 13 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |