Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACTTTGCCTGGATGTATTACCACA[C/T]GGGCAGACTGGCAGAAGCCCAGACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 147690 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IFIT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IFIT1 - interferon induced protein with tetratricopeptide repeats 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270927.1 | 599 | Missense Mutation | ACG,ATG | T,M 108 | NP_001257856.1 | |
NM_001270928.1 | 599 | Missense Mutation | ACG,ATG | T,M 77 | NP_001257857.1 | |
NM_001270929.1 | 599 | Missense Mutation | ACG,ATG | T,M 77 | NP_001257858.1 | |
NM_001270930.1 | 599 | Missense Mutation | ACG,ATG | T,M 77 | NP_001257859.1 | |
NM_001548.4 | 599 | Missense Mutation | ACG,ATG | T,M 108 | NP_001539.3 |