Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTTCCTAGTGGTGAAACTTTTGC[C/T]GATGCAGGTGTGTGCTAGTTCCTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616464 MIM: 609233 | ||||||||||||||||||||
Literature Links: |
ECD PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ECD - ecdysoneless cell cycle regulator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135752.1 | 2017 | Missense Mutation | AGC,GGC | S,G 592 | NP_001129224.1 | |
NM_001135753.1 | 2017 | Missense Mutation | AGC,GGC | S,G 516 | NP_001129225.1 | |
NM_007265.2 | 2017 | Missense Mutation | AGC,GGC | S,G 559 | NP_009196.1 |
NUDT13 - nudix hydrolase 13 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |