Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCACCACTCCCAGCTCGTTCTCGG[C/T]GGTGGTTAGCAGGTAGTTGGACTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606493 MIM: 172250 MIM: 616750 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EXOSC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EXOSC1 - exosome component 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318362.1 | 379 | Missense Mutation | ACC,GCC | T,A 122 | NP_001305291.1 | |
NM_001318363.1 | 379 | Intron | NP_001305292.1 | |||
NM_001318364.1 | 379 | Missense Mutation | ACC,GCC | T,A 81 | NP_001305293.1 | |
NM_001318365.1 | 379 | Missense Mutation | ACC,GCC | T,A 61 | NP_001305294.1 | |
NM_001318366.1 | 379 | Missense Mutation | ACC,GCC | T,A 61 | NP_001305295.1 | |
NM_016046.4 | 379 | Missense Mutation | ACC,GCC | T,A 147 | NP_057130.1 | |
XM_011539847.2 | 379 | Missense Mutation | ACC,GCC | T,A 112 | XP_011538149.1 | |
XM_017016304.1 | 379 | Missense Mutation | ACC,GCC | T,A 78 | XP_016871793.1 |
PGAM1 - phosphoglycerate mutase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZDHHC16 - zinc finger DHHC-type containing 16 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |