Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTCGACCACCTGGGAGGCTCCCTC[C/T]TGTGGCTGTTGGTCAAATGTGAACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608407 MIM: 611198 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DPYSL4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DPYSL4 - dihydropyrimidinase like 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006426.2 | 66 | Intron | NP_006417.2 | |||
XM_005252658.3 | 66 | UTR 5 | XP_005252715.1 | |||
XM_011539125.2 | 66 | Intron | XP_011537427.1 | |||
XM_017015482.1 | 66 | Intron | XP_016870971.1 | |||
XM_017015483.1 | 66 | Intron | XP_016870972.1 |
JAKMIP3 - Janus kinase and microtubule interacting protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001105521.3 | 66 | Intron | NP_001098991.1 | |||
NM_001323086.1 | 66 | Intron | NP_001310015.1 | |||
NM_001323087.1 | 66 | Intron | NP_001310016.1 | |||
NM_001323088.1 | 66 | Intron | NP_001310017.1 | |||
NM_001323089.1 | 66 | Intron | NP_001310018.1 | |||
NM_001323090.1 | 66 | Intron | NP_001310019.1 | |||
XM_005252674.4 | 66 | Intron | XP_005252731.1 | |||
XM_005252676.4 | 66 | Intron | XP_005252733.1 | |||
XM_011539676.2 | 66 | Intron | XP_011537978.1 | |||
XM_011539682.2 | 66 | Intron | XP_011537984.1 | |||
XM_017016095.1 | 66 | Intron | XP_016871584.1 | |||
XM_017016096.1 | 66 | Intron | XP_016871585.1 | |||
XM_017016097.1 | 66 | Intron | XP_016871586.1 | |||
XM_017016098.1 | 66 | Intron | XP_016871587.1 | |||
XM_017016099.1 | 66 | Intron | XP_016871588.1 |