Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATATGAAAAAAAATCAAAGCGAGT[A/G]TCATCTTTAGATACTTCTACTCATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610373 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DDX50 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DDX50 - DEAD-box helicase 50 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024045.1 | 908 | Silent Mutation | GTA,GTG | V,V 111 | NP_076950.1 | |
XM_005270148.2 | 908 | Silent Mutation | GTA,GTG | V,V 46 | XP_005270205.1 | |
XM_011540144.2 | 908 | Silent Mutation | GTA,GTG | V,V 46 | XP_011538446.1 | |
XM_017016626.1 | 908 | Silent Mutation | GTA,GTG | V,V 111 | XP_016872115.1 | |
XM_017016627.1 | 908 | Silent Mutation | GTA,GTG | V,V 46 | XP_016872116.1 | |
XM_017016628.1 | 908 | Silent Mutation | GTA,GTG | V,V 46 | XP_016872117.1 | |
XM_017016629.1 | 908 | Silent Mutation | GTA,GTG | V,V 46 | XP_016872118.1 | |
XM_017016630.1 | 908 | UTR 5 | XP_016872119.1 |