Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTGGGTTCAGAGAAAGAGGAGAA[G/T]AAAGGAAACCTCTGAGTGTCCATCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610734 MIM: 613597 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKRD2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANKRD2 - ankyrin repeat domain 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
C10orf62 - chromosome 10 open reading frame 62 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001009997.2 | 234 | Missense Mutation | AGA,ATA | R,I 10 | NP_001009997.2 |
HOGA1 - 4-hydroxy-2-oxoglutarate aldolase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134670.1 | 234 | Intron | NP_001128142.1 | |||
NM_138413.3 | 234 | Intron | NP_612422.2 |