Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGCTGCATTTCGCCCAGCAGCTCC[C/T]ATCACACCTCCTCCTGGAAGGGAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
PYROXD2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PYROXD2 - pyridine nucleotide-disulphide oxidoreductase domain 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032709.2 | 1778 | Missense Mutation | ATA,ATG | I,M 563 | NP_116098.2 | |
XM_011540293.2 | 1778 | Missense Mutation | ATA,ATG | I,M 597 | XP_011538595.1 | |
XM_011540301.2 | 1778 | Missense Mutation | ATA,ATG | I,M 294 | XP_011538603.1 | |
XM_017016835.1 | 1778 | UTR 3 | XP_016872324.1 | |||
XM_017016836.1 | 1778 | Intron | XP_016872325.1 | |||
XM_017016837.1 | 1778 | Intron | XP_016872326.1 | |||
XM_017016838.1 | 1778 | Missense Mutation | ATA,ATG | I,M 442 | XP_016872327.1 | |
XM_017016839.1 | 1778 | Missense Mutation | ATA,ATG | I,M 442 | XP_016872328.1 | |
XM_017016840.1 | 1778 | Missense Mutation | ATA,ATG | I,M 442 | XP_016872329.1 | |
XM_017016841.1 | 1778 | Missense Mutation | ATA,ATG | I,M 442 | XP_016872330.1 |