Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCATATCCTGTCTCCTTTTACTT[A/G]TGTTAATCATAGACTTTCGACATTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616753 MIM: 609767 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ENTPD7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ENTPD7 - ectonucleoside triphosphate diphosphohydrolase 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020354.3 | 308 | Missense Mutation | ATG,GTG | M,V 44 | NP_065087.1 |
SLC25A28 - solute carrier family 25 member 28 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |