Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACGGGAGTTGATGGGGATGACGC[A/C]GATGTCCACCACGACCACCACTCCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611380 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DIP2C PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DIP2C - disco interacting protein 2 homolog C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014974.2 | 2948 | Missense Mutation | NP_055789.1 | |||
XM_005252426.3 | 2948 | Missense Mutation | XP_005252483.1 | |||
XM_005252427.3 | 2948 | Missense Mutation | XP_005252484.1 | |||
XM_005252428.4 | 2948 | Missense Mutation | XP_005252485.1 | |||
XM_005252430.2 | 2948 | Missense Mutation | XP_005252487.2 | |||
XM_011519428.2 | 2948 | Missense Mutation | XP_011517730.1 | |||
XM_011519429.2 | 2948 | Missense Mutation | XP_011517731.1 | |||
XM_011519430.2 | 2948 | Missense Mutation | XP_011517732.1 | |||
XM_011519431.2 | 2948 | Missense Mutation | XP_011517733.1 | |||
XM_011519432.1 | 2948 | Missense Mutation | XP_011517734.1 | |||
XM_011519433.2 | 2948 | Missense Mutation | XP_011517735.1 | |||
XM_017015977.1 | 2948 | Missense Mutation | XP_016871466.1 |