Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCCGGAACAACAAGGAGGCGATGG[C/G]GCTCTACACCACCCATATCCTCAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602039 MIM: 608226 | ||||||||||||||||||||
Literature Links: |
EIF3A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EIF3A - eukaryotic translation initiation factor 3 subunit A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NANOS1 - nanos C2HC-type zinc finger 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_199461.3 | 760 | Missense Mutation | GCG,GGG | A,G 225 | NP_955631.1 |