Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCATCCTTGGACTGCGGGGCTGCAG[C/G]GGTGCAGCTGGGGGCTTTCCTGCGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609502 MIM: 616103 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDHR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CDHR1 - cadherin related family member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LRIT1 - leucine rich repeat, Ig-like and transmembrane domains 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LRIT2 - leucine rich repeat, Ig-like and transmembrane domains 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017924.3 | 1600 | Missense Mutation | CCT,GCT | P,A 511 | NP_001017924.1 | |
NM_001284223.1 | 1600 | Missense Mutation | CCT,GCT | P,A 521 | NP_001271152.1 |