Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCCTTGGAAGCCAATGACCCATTT[C/G]GTGAGTGAGGCCCCCAAACAGCCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616926 | ||||||||||||||||||||
Literature Links: |
FXYD4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FXYD4 - FXYD domain containing ion transport regulator 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001184963.1 | 402 | Missense Mutation | CCC,GCC | P,A 24 | NP_001171892.1 | |
NM_173160.2 | 402 | Missense Mutation | CCC,GCC | P,A 24 | NP_775183.1 |