Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTTCTCTGCAGGCCCCATGGGTC[C/T]ACCTGGAGAAATGCCATGTCCTCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 178630 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SFTPA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SFTPA1 - surfactant protein A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001093770.2 | Intron | NP_001087239.2 | ||||
NM_001164644.1 | Intron | NP_001158116.1 | ||||
NM_001164645.1 | Intron | NP_001158117.1 | ||||
NM_001164646.1 | Intron | NP_001158118.1 | ||||
NM_001164647.1 | Intron | NP_001158119.1 | ||||
NM_005411.4 | Intron | NP_005402.3 | ||||
XM_005270062.4 | Intron | XP_005270119.1 | ||||
XM_006717953.2 | Intron | XP_006718016.1 |