Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGACTGCTGTCGGTGTCGCGGCCG[C/G]AGCGGCCGCCCGAGTCGGCGGCGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616109 MIM: 608786 MIM: 605385 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C11orf80 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C11orf80 - chromosome 11 open reading frame 80 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001302084.1 | 2022 | UTR 3 | NP_001289013.1 | |||
NM_024650.3 | 2022 | UTR 3 | NP_078926.3 |
PC - pyruvate carboxylase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RCE1 - Ras converting CAAX endopeptidase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001032279.1 | 2022 | UTR 5 | NP_001027450.1 | |||
NM_005133.2 | 2022 | Missense Mutation | CAG,GAG | Q,E 18 | NP_005124.1 |