Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGACCTATGCTGTTATTTAGCTCAA[G/A]CTGCTCAGAGAGAATTAGTTCTGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608103 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALG8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ALG8 - ALG8, alpha-1,3-glucosyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001007027.2 | Intron | NP_001007028.1 | ||||
NM_024079.4 | Intron | NP_076984.2 | ||||
XM_005274247.3 | Intron | XP_005274304.1 | ||||
XM_011545252.2 | Intron | XP_011543554.1 | ||||
XM_017018274.1 | Intron | XP_016873763.1 |