Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGGTGCTGTGCCTGAGGAACAATA[C/T]CATTTTTAAGCAAGCCTTTTCTCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611853 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MRPL48 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MRPL48 - mitochondrial ribosomal protein L48 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318498.1 | 144 | UTR 5 | NP_001305427.1 | |||
NM_001318499.1 | 144 | Missense Mutation | ACC,ATC | T,I 15 | NP_001305428.1 | |
NM_001318500.1 | 144 | Missense Mutation | ACC,ATC | T,I 15 | NP_001305429.1 | |
NM_016055.5 | 144 | Missense Mutation | ACC,ATC | T,I 15 | NP_057139.1 | |
XM_017017919.1 | 144 | UTR 5 | XP_016873408.1 |