Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAACCCGTTACCCCAGAGACTCACC[C/T]CACCCTGCATAGTTGGTCAAACACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604626 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ME3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ME3 - malic enzyme 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001014811.1 | Intron | NP_001014811.1 | ||||
NM_001161586.1 | Intron | NP_001155058.1 | ||||
NM_006680.2 | Intron | NP_006671.2 | ||||
XM_005273717.1 | Intron | XP_005273774.1 | ||||
XM_017017136.1 | Intron | XP_016872625.1 | ||||
XM_017017137.1 | Intron | XP_016872626.1 | ||||
XM_017017138.1 | Intron | XP_016872627.1 |