Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTAAACTAATAGTTTAAGGAAAATA[C/T]AGGAAATGGGGGCAGGAGTAAAGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605390 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LPXN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LPXN - leupaxin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143995.2 | 1181 | Intron | NP_001137467.1 | |||
NM_001307951.1 | 1181 | Intron | NP_001294880.1 | |||
NM_004811.2 | 1181 | Intron | NP_004802.1 | |||
XM_006718750.1 | 1181 | Intron | XP_006718813.1 | |||
XM_011545388.2 | 1181 | Intron | XP_011543690.1 | |||
XM_011545390.2 | 1181 | Intron | XP_011543692.1 | |||
XM_011545391.2 | 1181 | Intron | XP_011543693.1 | |||
XM_011545393.2 | 1181 | UTR 3 | XP_011543695.1 | |||
XM_011545394.2 | 1181 | UTR 3 | XP_011543696.1 | |||
XM_017018579.1 | 1181 | Intron | XP_016874068.1 |