Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTCTGCGCCCCTTCCAACGCCTC[A/G]TCGTTGTCCTCTTCGTCCCCTGGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C11orf87 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C11orf87 - chromosome 11 open reading frame 87 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_207645.3 | 948 | Silent Mutation | TCA,TCG | S,S 147 | NP_997528.2 | |
XM_011542817.2 | 948 | Silent Mutation | TCA,TCG | S,S 294 | XP_011541119.1 | |
XM_011542818.2 | 948 | Silent Mutation | TCA,TCG | S,S 147 | XP_011541120.1 |