Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCTCAGTGCCATTAAATATATTA[C/T]CTTCTATTTCTTCTTAAAGAACTAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600814 | ||||||||||||||||||||
Literature Links: |
MRE11A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MRE11A - MRE11 homolog A, double strand break repair nuclease | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005590.3 | 2028 | Silent Mutation | AGA,AGG | R,R 680 | NP_005581.2 | |
NM_005591.3 | 2028 | Silent Mutation | AGA,AGG | R,R 708 | NP_005582.1 | |
XM_005274008.3 | 2028 | Silent Mutation | AGA,AGG | R,R 552 | XP_005274065.1 | |
XM_006718842.3 | 2028 | Silent Mutation | AGA,AGG | R,R 707 | XP_006718905.1 | |
XM_011542837.2 | 2028 | Silent Mutation | AGA,AGG | R,R 708 | XP_011541139.1 | |
XM_017017772.1 | 2028 | Silent Mutation | AGA,AGG | R,R 708 | XP_016873261.1 | |
XM_017017773.1 | 2028 | Silent Mutation | AGA,AGG | R,R 707 | XP_016873262.1 |