Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGCTGGACAAGTTCTCAAAGAAG[C/G]TGCAGCCGGAGCCAAACAGTGTCAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ATG16L2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ATG16L2 - autophagy related 16 like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318766.1 | 188 | UTR 5 | NP_001305695.1 | |||
NM_033388.1 | 188 | Missense Mutation | CTG,GTG | L,V 57 | NP_203746.1 | |
XM_005274376.4 | 188 | Missense Mutation | CTG,GTG | L,V 57 | XP_005274433.1 | |
XM_006718732.2 | 188 | Missense Mutation | CTG,GTG | L,V 57 | XP_006718795.1 | |
XM_006718733.3 | 188 | Missense Mutation | CTG,GTG | L,V 57 | XP_006718796.1 | |
XM_006718734.2 | 188 | Intron | XP_006718797.1 | |||
XM_011545332.1 | 188 | Missense Mutation | CTG,GTG | L,V 57 | XP_011543634.1 | |
XM_011545333.1 | 188 | UTR 5 | XP_011543635.1 | |||
XM_011545334.1 | 188 | UTR 5 | XP_011543636.1 |