Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTTGATTGTCAGGCGCATCATTTC[A/G]AAGTGGCCCCAATGCAGGTAGCCAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611829 MIM: 600876 | ||||||||||||||||||||
Literature Links: |
MRPL16 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MRPL16 - mitochondrial ribosomal protein L16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017840.3 | 520 | Silent Mutation | TTC,TTT | F,F 102 | NP_060310.1 |
STX3 - syntaxin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001178040.1 | 520 | Intron | NP_001171511.1 | |||
NM_004177.4 | 520 | Intron | NP_004168.1 | |||
XM_005274195.3 | 520 | Intron | XP_005274252.1 | |||
XM_005274198.2 | 520 | Intron | XP_005274255.1 | |||
XM_005274200.3 | 520 | Intron | XP_005274257.1 | |||
XM_011545221.2 | 520 | Intron | XP_011543523.1 | |||
XM_017018188.1 | 520 | Intron | XP_016873677.1 | |||
XM_017018189.1 | 520 | Intron | XP_016873678.1 | |||
XM_017018190.1 | 520 | Intron | XP_016873679.1 | |||
XM_017018191.1 | 520 | Intron | XP_016873680.1 | |||
XM_017018192.1 | 520 | Intron | XP_016873681.1 | |||
XM_017018193.1 | 520 | Intron | XP_016873682.1 | |||
XM_017018194.1 | 520 | Intron | XP_016873683.1 | |||
XM_017018195.1 | 520 | Intron | XP_016873684.1 | |||
XM_017018196.1 | 520 | Intron | XP_016873685.1 |