Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAATCAATACTGTAGAACACTCACA[C/T]GCTGATTTATCACTAGGCACAAATC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602158 | ||||||||||||||||||||
Literature Links: |
CLNS1A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CLNS1A - chloride nucleotide-sensitive channel 1A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001293.2 | 455 | Intron | NP_001284.1 | |||
NM_001311199.1 | 455 | Silent Mutation | GCA,GCG | A,A 121 | NP_001298128.1 | |
NM_001311200.1 | 455 | Intron | NP_001298129.1 | |||
NM_001311201.1 | 455 | Intron | NP_001298130.1 | |||
NM_001311202.1 | 455 | Intron | NP_001298131.1 |
LOC107987163 - uncharacterized LOC107987163 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |