Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602141 MIM: 604592 | ||||||||||||||||||||
Literature Links: |
MIR4691 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MIR4691 - microRNA 4691 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR6753 - microRNA 6753 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR7113 - microRNA 7113 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NDUFS8 - NADH:ubiquinone oxidoreductase core subunit S8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TCIRG1 - T-cell immune regulator 1, ATPase H+ transporting V0 subunit a3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006019.3 | 206 | Missense Mutation | CGG,TGG | R,W 28 | NP_006010.2 | |
NM_006053.3 | 206 | Intron | NP_006044.1 | |||
XM_005273709.3 | 206 | Missense Mutation | CGG,TGG | R,W 28 | XP_005273766.1 | |
XM_011544726.2 | 206 | Missense Mutation | CGG,TGG | R,W 28 | XP_011543028.1 | |
XM_017017089.1 | 206 | Missense Mutation | CGG,TGG | R,W 28 | XP_016872578.1 | |
XM_017017090.1 | 206 | UTR 5 | XP_016872579.1 |