Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTCCCCATAGCGGGTAACTTGGGA[C/T]GGTGGGTACTTGGCCAGCCAGGCAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602289 | ||||||||||||||||||||
Literature Links: |
C11orf68 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C11orf68 - chromosome 11 open reading frame 68 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135635.1 | 329 | Silent Mutation | CCA,CCG | P,P 101 | NP_001129107.1 | |
NM_031450.3 | 329 | Silent Mutation | CCA,CCG | P,P 100 | NP_113638.2 |
DRAP1 - DR1 associated protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |