Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAACACATGAACAGAAGACGAAAA[C/T]TTCTTCTAGCCTCAGTACTTGCTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
DDIAS PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DDIAS - DNA damage induced apoptosis suppressor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145018.3 | 125 | Missense Mutation | CTT,TTT | L,F 7 | NP_659455.3 | |
XM_011544835.1 | 125 | Missense Mutation | CTT,TTT | L,F 7 | XP_011543137.1 | |
XM_011544836.2 | 125 | Missense Mutation | CTT,TTT | L,F 7 | XP_011543138.1 | |
XM_011544837.1 | 125 | Intron | XP_011543139.1 | |||
XM_011544838.2 | 125 | UTR 5 | XP_011543140.1 | |||
XM_017017352.1 | 125 | UTR 5 | XP_016872841.1 | |||
XM_017017353.1 | 125 | UTR 5 | XP_016872842.1 |