Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCCGGCAGCCAATGTAGCCGTCCC[C/G]GTCTCGGTCAAACTCCTGGAAGGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607314 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CABP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CABP2 - calcium binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318496.1 | 386 | Missense Mutation | CGG,GGG | R,G 100 | NP_001305425.1 | |
NM_016366.2 | 386 | Missense Mutation | CGG,GGG | R,G 94 | NP_057450.2 |