Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGAGTGGGAAGGTATCCCCTTTCC[A/G]TAGGCTCAGTCCTCTCCAGGCCCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
9 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606203 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GAB2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GAB2 - GRB2 associated binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012296.3 | 5076 | UTR 3 | NP_036428.1 | |||
NM_080491.2 | 5076 | UTR 3 | NP_536739.1 | |||
XM_006718753.2 | 5076 | UTR 3 | XP_006718816.1 | |||
XM_011545408.2 | 5076 | UTR 3 | XP_011543710.1 |
USP35 - ubiquitin specific peptidase 35 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020798.2 | 5076 | Intron | NP_065849.1 | |||
XM_011545187.2 | 5076 | Intron | XP_011543489.1 | |||
XM_011545188.2 | 5076 | Intron | XP_011543490.1 | |||
XM_017018052.1 | 5076 | Intron | XP_016873541.1 | |||
XM_017018053.1 | 5076 | Intron | XP_016873542.1 |