Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGAGTTCGCTTAGCTTGTGTCGAC[A/G]CACTTCACAAGCATGGAGGAGTTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608210 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EXT2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EXT2 - exostosin glycosyltransferase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000401.3 | Intron | NP_000392.3 | ||||
NM_001178083.1 | Intron | NP_001171554.1 | ||||
NM_207122.1 | Intron | NP_997005.1 | ||||
XM_011519950.1 | Intron | XP_011518252.1 | ||||
XM_011519951.1 | Intron | XP_011518253.1 |