Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCCCCATGCCCGGGAAGTGAAACA[C/T]GTGCATTTAGAAAACGCTACAGAGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610493 | ||||||||||||||||||||
Literature Links: |
C11orf52 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C11orf52 - chromosome 11 open reading frame 52 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_080659.2 | 378 | Silent Mutation | CAC,CAT | H,H 94 | NP_542390.2 |
DIXDC1 - DIX domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HSPB2-C11orf52 - HSPB2-C11orf52 readthrough (NMD candidate) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |